2,6-Lutidine

Synonyms: 2,6-Dimethylpyridine

336106-2L 336106-100ML 336106-1L
220057-780EA 299.08 USD
220057-780 220057-778 220057-779
2,6-Lutidine
2,6-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 143.7 °C (1013 hPa)
Melting Pt: –6 °C
Density: 0.9252 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00006345
CAS Number: 108-48-5
EINECS: 203-587-3
UN: 1993
ADR: 3,III
Merck Index: 13,05636

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR