2,5-Dimethyl-p-phenylenediamine ≥97%

Synonyms: 2,5-Diamino-p-xylene, 2,5-Dimethyl-1,4-phenylenediamine, 1,4-Diamino-2,5-dimethylbenzene, 2,5-Dimethylbenzene-1,4-diamine

336068-5G 336068-25G
220056-953EA 160.85 USD
220056-953 220056-952
2,5-Dimethyl-p-phenylenediamine ≥97%
2,5-Dimethyl-p-phenylenediamine
Formula: C₈H₁₂N₂
MW: 136.2 g/mol
MDL Number: MFCD00052675
CAS Number: 6393-01-7
EINECS: 229-003-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR