β-Chloropropiophenone ≥98%

Synonyms: 3-Chloropropiophenone

335614-10G 335614-50G
220057-460EA 89.17 USD
220057-460 220057-461
β-Chloropropiophenone ≥98%
β-Chloropropiophenone
Formula: (C₆H₅)C(O)CH₂CH₂Cl
MW: 168.62 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 48…49 °C
Density: 1.181 g/cm³ (20 °C)
MDL Number: MFCD00000990
CAS Number: 936-59-4
EINECS: 213-317-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR