5-Methoxy-o-toluidine ≥97%

Synonyms: 2-Amino-4-methoxytoluene, 3-Amino-4-methylanisole, 1-Amino-3-methoxy-6-methylbenzene, 6-Methyl-m-anisidine, 5-Methoxy-2-methylaniline

335436-5G 335436-1G
220057-058EA 239.26 USD
220057-058 220057-057
5-Methoxy-o-toluidine ≥97%
5-Methoxy-o-toluidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 47 °C
Density: 1.39 g/cm³ (25 °C)
MDL Number: MFCD00075057
CAS Number: 50868-72-9
EINECS: 256-816-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR