3-Aminophthalic acid ≥90%, Technical Grade

335347-1G 335347-5G
220057-038EA 165 USD
220057-038 220057-039
3-Aminophthalic acid ≥90%, Technical Grade
3-Aminophthalic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
CAS Number: 5434-20-8
EINECS: 226-595-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR