Tetramethylammonium hydroxide 25% (w/w) in methanol

334901-500ML 334901-100ML
220056-944EA 99.23 USD
220056-944 220056-943
Tetramethylammonium hydroxide 25% (w/w) in methanol
Tetramethylammonium hydroxide
Formula: C₄H₁₃NO
MW: 91.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR