4-Chloro-2-fluorophenol ≥99%

334227-5G 334227-25G
220057-028EA 42.97 USD
220057-028 220057-027
4-Chloro-2-fluorophenol ≥99%
4-Chloro-2-fluorophenol
Formula: C₆H₄ClFO
MW: 146.55 g/mol
MDL Number: MFCD00075030
CAS Number: 348-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR