3-Amino-2-methylbenzyl alcohol ≥97%

Synonyms: (3-Amino-2-methylphenyl)methanol

334200-10G 334200-50G
220057-025EA 222.31 USD
220057-025 220057-026
3-Amino-2-methylbenzyl alcohol ≥97%
3-Amino-2-methylbenzyl alcohol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00075028
CAS Number: 83647-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR