Tetraethyl orthosilicate (TEOS) ≥99.999%

Danger

Synonyms: Tetraethoxysilane

333859-25ML 333859-100ML
220057-527EA 83.69 USD
220057-527 220057-526
Tetraethyl orthosilicate (TEOS) ≥99.999%
Tetraethyl orthosilicate (TEOS)
Formula: Si(OC₂H₅)₄
MW: 208.33 g/mol
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: -85.5 °C
Density: 0.933 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00009062
CAS Number: 78-10-4
EINECS: 201-083-8
UN: 1292
ADR: 3,III
Merck Index: 13,03882

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR