2,3,4-Trifluorobenzoic acid ≥98%

Warning

333824-25G 333824-5G
220057-520EA 69.46 USD
220057-520 220057-521
2,3,4-Trifluorobenzoic acid ≥98%
2,3,4-Trifluorobenzoic acid
Formula: C₇H₃F₃O₂
MW: 176.09 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 140 - 142 °C
MDL Number: MFCD00061232
CAS Number: 61079-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR