Iridium(III) acetylacetonate

Synonyms: Iridium(III) 2,4-acetylacetonate, Tris(2,4-pentanedionato-O,O')iridium (III), Iridium (III) tris(2,4-pentanedionato-O,O'), Iridium (III) 2,4-pentanedionate

333352-500MG 333352-2.5G
220057-366EA 335.38 USD
220057-366 220057-365
Iridium(III) acetylacetonate
Iridium(III) acetylacetonate
Formula: C₁₅H₂₁IrO₆
MW: 489.55 g/mol
MDL Number: MFCD00015353
CAS Number: 15635-87-7
EINECS: 239-711-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR