Titanium(IV) fluoride

333239-10G 333239-100G
220057-357EA 45 USD
220057-357 220057-356
Titanium(IV) fluoride
Titanium(IV) fluoride
Formula: F₄Ti
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 400 °C
Density: 2.798 g/cm³ (20.5 °C)
MDL Number: MFCD00042545
CAS Number: 7783-63-3
EINECS: 232-017-6
UN: 3260
ADR: 8,II
Merck Index: 12,09619

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR