Lanthanum(III) nitrate hexahydrate ≥99.99%

Warning

331937-500G 331937-100G 331937-5G
220056-749EA 936.92 USD
220056-749 220056-748 220056-750
Lanthanum(III) nitrate hexahydrate ≥99.99%
Lanthanum(III) nitrate hexahydrate
Formula: La(NO₃)₃·6H₂O
MW: 433.01 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: 65 - 67 °C
Density: 2.347 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149751
CAS Number: 10277-43-7
EINECS: 233-238-0
UN: 1477
ADR: 5.1,III
Merck Index: 14,05363

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR