trans-4-Methylcyclohexanecarboxylic acid ≥98%

Synonyms: trans-4-Methyl-1-cyclohexanecarboxylic acid

331600-5G 331600-1G
220057-188EA 30.15 USD
220057-188 220056-702
trans-4-Methylcyclohexanecarboxylic acid ≥98%
trans-4-Methylcyclohexanecarboxylic acid
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
MDL Number: MFCD00074943
CAS Number: 13064-83-0
EINECS: 235-959-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR