2,5-Diiodothiophene

331570-5G 331570-1G
220055-811EA 114.15 USD
220055-811 220055-810
2,5-Diiodothiophene
2,5-Diiodothiophene
Formula: C₄H₂I₂S
MW: 335.93 g/mol
Boiling Pt: 67…70 °C (0.05 mmHg)
Melting Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014525
CAS Number: 625-88-7
EINECS: 210-915-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR