Yttrium(III) nitrate tetrahydrate

Warning

331309-100G 331309-10G
220057-168EA 612.31 USD
220057-168 220057-169
Yttrium(III) nitrate tetrahydrate
Yttrium(III) nitrate tetrahydrate
Formula: N₃O₉Y·4H₂O
MW: 346.98 g/mol
MDL Number: MFCD00149938
CAS Number: 13773-69-8
EINECS: 233-802-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR