Perfluoro-tert-butanol ≥99%

Synonyms: Perfluoro-t-butanol, Nonafluoro-tert-butyl alcohol, 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol, Perfluoro-tert-butyl alcohol

331023-5G 331023-1G
220056-913EA 187.38 USD
220056-913 220056-912
Perfluoro-tert-butanol ≥99%
Perfluoro-tert-butanol
Formula: C₄HF₉O
MW: 236.04 g/mol
Boiling Pt: 84 °C (1013 hPa)
Melting Pt: –17 °C
Density: 1.693 g/cm³ (20 °C)
MDL Number: MFCD00042092
CAS Number: 2378-02-1
EINECS: 219-157-3
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR