Titanyl(IV) acetylacetonate

Synonyms: Titanium(IV) oxide acetylacetonate, Titanium(IV) oxyacetylacetonate, Titanyl Acetylacetonate, Titanium (IV) oxide bis(acetylacetonate), Oxobis(2,4-pentanedionato-O,O')titanium (IV), Titanium (IV) oxide bis(2,4-pentanedionate)

330833-50G 330833-10G
220057-089EA 429.23 USD
220057-089 220057-088
Titanyl(IV) acetylacetonate
Titanyl(IV) acetylacetonate
Formula: C₁₀H₁₄O₅Ti
MW: 262.1 g/mol
MDL Number: MFCD00013505
CAS Number: 14024-64-7
EINECS: 237-861-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR