Cesium acetate ≥99.9%

Synonyms: Acetic acid cesium salt

329827-25G 329827-100G
220056-874EA 130.15 USD
220056-874 220056-873
Cesium acetate ≥99.9%
Cesium acetate
Formula: H₃CCOOCs
MW: 191.95 g/mol
Melting Pt: 194 °C
MDL Number: MFCD00013056
CAS Number: 3396-11-0
EINECS: 222-248-0

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR