Ethyl-3-(trimethylsilyl)propiolate ≥99%

Synonyms: Ethyl 3-(trimethylsilyl)propynoate, 3-(Trimethylsilyl)-2-propynoic acid ethyl ester, 3-(Trimethylsilyl)propiolic acid ethyl ester, Ethyl 3-(trimethylsilyl)propiolate

329452-25G 329452-5G
220056-839EA 288.62 USD
220056-839 220056-840
Ethyl-3-(trimethylsilyl)propiolate ≥99%
Ethyl-3-(trimethylsilyl)propiolate
Formula: C₈H₁₄O₂Si
MW: 170.28 g/mol
MDL Number: MFCD00074886
CAS Number: 16205-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR