Charcoal activated, pellets, Norit® ROW 0.8 SUPRA, Sigma-Aldrich®

329428-2KG 329428-500G
220056-833EA 231.54 USD
220056-833 220056-834
Charcoal activated, pellets, Norit® ROW 0.8 SUPRA, Sigma-Aldrich®
Charcoal activated
Formula: C
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR