4-Fluoro-3-nitrobenzoic acid ≥98%

Warning

329045-5G 329045-25G
220056-801EA 28 USD
220056-801 220056-800
4-Fluoro-3-nitrobenzoic acid ≥98%
4-Fluoro-3-nitrobenzoic acid
Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Boiling Pt: 92 °C (15 -rr)
Melting Pt: 123 - 124 °C
MDL Number: MFCD00007058
CAS Number: 453-71-4
EINECS: 207-221-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR