Bromocresol purple sodium salt indicator

Synonyms: Sodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonate, Bromocresol purple water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolate

860891-25G 860891-5G
220098-062EA 148.4 USD
220098-062 220098-063
Bromocresol purple sodium salt indicator
Bromocresol purple sodium salt
Formula: C₂₁H₁₅Br₂NaO₅S
MW: 562.21 g/mol
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00148896
CAS Number: 62625-30-3
EINECS: 263-655-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ae99d0ddad367ac12f93884d5b8f055a043d863&vid=d52d7764c82f6e9c25fd1b36b6fd236547dd8c8c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR