Dimethyl diselenide ≥96%

Danger

Synonyms: Dimethyldiselenide

328502-1G 328502-5G
220056-617EA 98.86 USD
220056-617 220056-618
Dimethyl diselenide ≥96%
Dimethyl diselenide
Formula: C₂H₆Se₂
MW: 187.99 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –82 °C
Density: 2.0009 g/cm³ (20 °C)
MDL Number: MFCD00008254
CAS Number: 7101-31-7
EINECS: 230-410-7
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR