Tetramethylammonium hydroxide 10% (w/w) in aqueous solution

Danger

328251-250ML 328251-1L
220056-579EA 52.92 USD
220056-579 220056-578
Tetramethylammonium hydroxide 10% (w/w) in aqueous solution
Tetramethylammonium hydroxide
Formula: C₄H₁₃NO
MW: 91.15 g/mol
Boiling Pt: 110 °C (1013 hPa)
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR