Palladium ≥99.9% (trace metals basis), powder <1 µm

Danger

326666-1G 326666-5G
220056-018EA 284.62 USD
220056-018 220056-019
Palladium ≥99.9% (trace metals basis), powder <1 µm
Palladium
Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR