Indium(III) nitrate hydrate ≥99.9%

326135-50G 326135-10G
220056-433EA 424.62 USD
220056-433 220056-432
Indium(III) nitrate hydrate ≥99.9%
Indium(III) nitrate hydrate
Formula: InN₃O₉·H₂O
MW: 318.85 g/mol
MDL Number: MFCD00149742
CAS Number: 207398-97-8
EINECS: 237-393-5
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR