Indium(III) nitrate hydrate ≥99.99%

326127-10G 326127-50G
220056-429EA 156.92 USD
220056-429 220056-430
Indium(III) nitrate hydrate ≥99.99%
Indium(III) nitrate hydrate
Formula: InN₃O₉·H₂O
MW: 318.85 g/mol
MDL Number: MFCD00149742
CAS Number: 207398-97-8
EINECS: 237-393-5
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR