Ytterbium triacetate tetrahydrate ≥99.9%

Synonyms: Ytterbium(III) acetate tetrahydrate

326011-50G 326011-10G
220056-420EA 416.92 USD
220056-420 220056-419
Ytterbium triacetate tetrahydrate ≥99.9%
Ytterbium triacetate tetrahydrate
Formula: C₆H₉O₆Yb·4H₂O
MW: 422.23 g/mol
CAS Number: 15280-58-7
EINECS: 244-137-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR