Terbium trinitrate pentahydrate ≥99.9%

Synonyms: Terbium(III) nitrate hydrate, Terbium (III) nitrate pentahydrate

325945-25G 325945-5G
220056-409EA 483.08 USD
220056-409 220056-410
Terbium trinitrate pentahydrate ≥99.9%
Terbium trinitrate pentahydrate
Formula: N₃O₉Tb·5H₂O
MW: 435.02 g/mol
Melting Pt: 89.3 °C
MDL Number: MFCD00149867
CAS Number: 57584-27-7
EINECS: 233-138-7
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR