Terbium triacetate hydrate ≥99.9%

Synonyms: Terbium(III) acetate hydrate

325929-25G 325929-5G
220056-405EA 926.15 USD
220056-405 220056-406
Terbium triacetate hydrate ≥99.9%
Terbium triacetate hydrate
Formula: C₆H₉O₆Tb·H₂O
MW: 354.07 g/mol
MDL Number: MFCD00149233
CAS Number: 100587-92-6
EINECS: 247-065-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR