Scandium(III) nitrate hydate

Synonyms: Trinitric acid scandium salt hydrate, Scandium(3+) trinitrate hydrate, Scandium trinitrate hydrate

325902-1G 325902-5G
220056-400EA 252.31 USD
220056-400 220056-401
Scandium(III) nitrate hydate
Scandium(III) nitrate hydrate
Formula: Sc(NO₃)₃·xH₂O
MW: 230.97 g/mol (anhy)
MDL Number: MFCD00011220
CAS Number: 13465-60-6
EINECS: 236-701-5
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR