Holmium trinitrate pentahydrate ≥99.9%

Danger

325732-10G 325732-50G
220055-902EA 215.38 USD
220055-902 220055-903
Holmium trinitrate pentahydrate ≥99.9%
Holmium trinitrate pentahydrate
Formula: HoN₃O₉·5H₂O
MW: 441.02 g/mol
Melting Pt: 83.74…84.62 °C
MDL Number: MFCD00149737
CAS Number: 14483-18-2
EINECS: 233-438-8
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR