Europium triacetate hydrate ≥99.9%

Synonyms: Acetic acid europium (III) salt hydrate, Europium(III) acetate xhydrate, Europium (III) acetate hydrate

325627-25G 325627-5G
220056-394EA 1167.69 USD
220056-394 220056-395
Europium triacetate hydrate ≥99.9%
Europium triacetate hydrate
Formula: C₆H₉EuO₆·H₂O
MW: 347.11 g/mol
MDL Number: MFCD00150115
CAS Number: 62667-64-5
EINECS: 214-670-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR