Erbium triacetate hydrate ≥99.9%

Synonyms: Erbium(III) acetate hydrate

325570-10G 325570-50G
220055-849EA 134.15 USD
220055-849 220056-389
Erbium triacetate hydrate ≥99.9%
Erbium triacetate hydrate
Formula: C₆H₉ErO₆·H₂O
CAS Number: 304675-52-3
EINECS: 247-067-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR