2-Methylnicotinic acid

Synonyms: 2-Methylpyridine-3-carboxylic acid, 2-Methyl-3-pyridinecarboxylic acid

325228-5G 325228-1G
220056-304EA 263.85 USD
220056-304 220056-303
2-Methylnicotinic acid
2-Methylnicotinic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD00013449
CAS Number: 3222-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR