3-Nitrophenylboronic acid

Synonyms: (3-Nitrophenyl)metaboric acid, Dihydroxy(3-nitrophenyl)borane, 3-Nitrobenzeneboronic acid

325104-5G 325104-1G
220056-286EA 46.31 USD
220056-286 220056-285
3-Nitrophenylboronic acid
3-Nitrophenylboronic acid
Formula: C₆H₆BNO₄
MW: 166.93 g/mol
Melting Pt: 290…291 °C
MDL Number: MFCD00007193
CAS Number: 13331-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR