2-Bromo-5-nitropyridine

Warning

324833-5G 324833-1G
220056-254EA 31.02 USD
220056-254 220056-253
2-Bromo-5-nitropyridine
2-Bromo-5-nitropyridine
Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
MDL Number: MFCD00006222
CAS Number: 4487-59-6
EINECS: 224-777-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR