2,2,2-Trifluorothanesulfonyl chloride ≥99%

Synonyms: 2,2,2-Trifluoroethanesulphonyl chloride, (2,2,2-Trifluoroethyl)sulphonyl chloride

324787-1G 324787-250MG 324787-5G
220056-246EA 306.92 USD
220056-246 220056-247 220056-248
2,2,2-Trifluorothanesulfonyl chloride ≥99%
2,2,2-Trifluorothanesulphonyl chloride
Formula: C₂H₂ClF₃O₂S
MW: 182.55 g/mol
Boiling Pt: 135 °C (1013 hPa)
Density: 1.6905 g/cm³ (23 °C)
MDL Number: MFCD00007458
CAS Number: 1648-99-3
EINECS: 216-713-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR