Ethyl-6-bromohexanoate ≥99%

324728-100G 324728-25G
220056-240EA 266 USD
220056-240 220056-241
Ethyl-6-bromohexanoate ≥99%
Ethyl-6-bromohexanoate
Formula: BrH₂C(CH₂)₄COOCH₂CH₃
Boiling Pt: 69…72 °C (0.1 mmHg)
Melting Pt: 33 °C
Density: 1.2592 g/cm³ (20 °C)
MDL Number: MFCD00000270
CAS Number: 25542-62-5
EINECS: 247-085-2

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR