Lithiumbis(trimethylsilyl)amide

Synonyms: Lithiumbis(triméthylsilyl)amide, HMDS-Li, N-Lithiohexamethyldisilazane, HMDS lithium salt

324620-50G 324620-10G 324620-250G
220056-229EA 88.62 USD
220056-229 220056-227 220056-228
Lithiumbis(trimethylsilyl)amide
Lithiumbis(trimethylsilyl)amide
Formula: C₆H₁₈LiNSi₂
MW: 167.33 g/mol
Boiling Pt: 80…84 °C (0.01 mmHg)
Melting Pt: 105…107 °C
Density: 0.89 g/cm³ (20 °C)
MDL Number: MFCD00008261
CAS Number: 4039-32-1
EINECS: 223-725-6
UN: 3396
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR