Potassium tetrachloroplatinate(II) ≥99.99%

Danger

Synonyms: Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate

323411-1G 323411-250MG 323411-5G
220056-093EA 306.15 USD
220056-093 220056-094 220056-095
Potassium tetrachloroplatinate(II) ≥99.99%
Potassium tetrachloroplatinate(II)
Formula: Cl₄K₂Pt
MW: 415.09 g/mol
Boiling Pt: 35 - 40 °C (20 -rr)
Melting Pt: 265 °C
Density: 3.38 g/cm³ (20 °C)
MDL Number: MFCD00011378
CAS Number: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1,III
Merck Index: 12,07855

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR