2-Nitrobenzoyl chloride ≥97%

322717-25G 322717-5G
220055-880EA 172.31 USD
220055-880 220055-881
2-Nitrobenzoyl chloride ≥97%
2-Nitrobenzoyl chloride
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 25 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007131
CAS Number: 610-14-0
EINECS: 210-206-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR