Diethylene glycol monobutyl ether ≥98.0% (by GC)

Warning

Synonyms: 2-(2-Butoxyethoxy)ethanol, Diethylene glycol mono-n-butyl ether, Butyldigol

32250-1L-F 32250-2.5L-F
220055-819EA 13.23 USD
220055-819 220055-820
Diethylene glycol monobutyl ether ≥98.0% (by GC)
Diethylene glycol monobutyl ether
Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MW: 162.23 g/mol
Boiling Pt: 230.4 °C (1013 hPa)
Melting Pt: –68 °C
Density: 0.954 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002881
CAS Number: 112-34-5
EINECS: 203-961-6
Merck Index: 13,01556

Order Now

Specification Test Results

Assay 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR