Tetrapotassium diphosphate ≥97%

Warning

Synonyms: Tetrapotassium diphosphate, Potassium diphosphate, Potassium pyrophosphate, tetra-Potassium pyrophosphate, Potassium pyrophospate

322431-500G 322431-25G 322431-2.5KG
220053-171EA 66.77 USD
220053-171 220053-170 220053-169
Tetrapotassium diphosphate ≥97%
Tetrapotassium diphosphate
Formula: K₄O₇P₂
MW: 330.34 g/mol
Boiling Pt: ∼1500 °C (1013 hPa)
Melting Pt: 1090 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00011393
CAS Number: 7320-34-5
EINECS: 230-785-7
Merck Index: 12,07833

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR