4-(Methylamino)phenol sulfate ≥99% ACS

Warning

Synonyms: p-(Methylamino)phenol sulphate, Metol, 4-Methylaminophenol sulphate, 4-(Methylamino)phenol hemisulphate, 4-Methylaminophenolsulphate, 4-(Methylamino)phenol hemisulphate salt

320013-100G 320013-500G
220055-609EA 53.31 USD
220055-609 220055-251
4-(Methylamino)phenol sulfate ≥99% ACS
4-(Methylamino)phenol sulphate
Formula: C₁₄H₂₀N₂O₆S
MW: 344.39 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.434 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013140
CAS Number: 55-55-0
EINECS: 200-237-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,06046

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR