trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98% ACS

Warning

Synonyms: Titriplex® IV

319945-100G 319945-25G
220054-795EA 153.08 USD
220054-795 220054-796
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98% ACS
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Formula: C₁₄H₂₂N₂O₈·1H₂O
MW: 364.35 g/mol
Melting Pt: 213…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00066429
CAS Number: 125572-95-4
EINECS: 236-308-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR