Mercury(II) nitrate 0.01 N in aqueous solution

Warning

Synonyms: Mercury dinitrate, Mercuric nitrate

319066-2L 319066-450ML
220055-498EA 232.31 USD
220055-498 220055-499
Mercury(II) nitrate 0.01 N in aqueous solution
Mercury(II) nitrate
Formula: HgN₂O₆
MW: 324.60 g/mol
MDL Number: MFCD00011038
CAS Number: 10045-94-0
EINECS: 233-152-3
UN: 3287
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR