Bromocresol purple 0.04% (w/w) in aqueous solution

Synonyms: 5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

318736-100ML 318736-500ML
220055-469EA 32.2 USD
220055-469 220055-470
Bromocresol purple 0.04% (w/w) in aqueous solution
Bromocresol purple
Formula: C₂₁H₁₆Br₂O₅S
MW: 540.23 g/mol
MDL Number: MFCD00011681
CAS Number: 115-40-2
EINECS: 204-087-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47c9670cec7df9e6d9e9df64f2a1265a398b2114&vid=cecd43940ef16211ee326132ea630de2e014f984&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR