1-Iodoperfluorbutane ≥98%

Synonyms: Perfluorobutyl iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, Nonafluoro-1-iodobutane

317845-100G 317845-25G
220055-137EA 188.46 USD
220055-137 220055-138
1-Iodoperfluorbutane ≥98%
1-Iodoperfluorbutane
Formula: C₄F₉I
MW: 345.93 g/mol
Boiling Pt: 65 °C (1013 hPa)
Melting Pt: –2 °C
Density: 2.0497 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001062
CAS Number: 423-39-2
EINECS: 207-025-8
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR